Order Entry
Singapore
ContactUsLinkComponent
bisbenzimide H-33342 trihydrochloride (Hoechst 33342), light yellow powder
bisbenzimide H-33342 trihydrochloride (Hoechst 33342), light yellow powder
Catalog # ICNA0219030525
Supplier:  MP Biomedicals
CAS Number:  
bisbenzimide H-33342 trihydrochloride (Hoechst 33342), light yellow powder
Catalog # ICNA0219030525
Supplier:  MP Biomedicals
CAS Number:  

Specifications

  • Pk:
    25 mg
  • Seq:
    2315

Specifications

Specification Test Results

  • Identity Test :
    Passes
  • λmax (methanol:
    352 ± 3 nm

Specification Test Results

About this item

 :
 :561.93 g/mol
Storage Temperature:Refrigerator
CAS Number:23491-52-3
 :MFCD00012679
Formula:C₂₇H₂₈N₆O·3HCl