Order Entry
Singapore
ContactUsLinkComponent
Calcium pantothenate USP
Calcium pantothenate USP
Catalog # ICNA0210122805
Supplier:  MP Biomedicals
CAS Number:  
Calcium pantothenate USP
Catalog # ICNA0210122805
Supplier:  MP Biomedicals
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Identity Test A:
    Positive
  • Identity Test B:
    Positive
  • Alkalinity:
    Passes
  • Specific Rotation (c=5, water):
    +25.0 to + 27.5°
  • Loss on Drying:
    ≤5.0%
  • Heavy Metals:
    ≤0.002%
  • Calcium Content (dry basis):
    8.2 - 8.6%
  • Nitrogen Content (dry basis):
    5.7 - 6.0%
  • Ordinary Impurities:
    ≤1.0%
  • Assay:
    98%-101%
  • Total Plate Count:
    <1000 cfu/g
  • Yeast & Mold:
    <100 cfu/g

Specification Test Results

About this item

 :
 :476.54 g/mol
Melting Pt:170…172 °C (lit.)
Storage Temperature:Ambient
CAS Number:137-08-6
Formula:C₁₈H₃₂CaN₂O₁₀
MDL Number:MFCD00002766