Horse Cytochrome C (from Heart), MP Biomedicals
Cytochrome C is an electron-carrying mitochondrial protein. The transition of Cytochrome C between the ferrous and ferric states within the cell makes it an efficient biological electron-transporter and it plays a vital role in cellular oxidations in both plants and animals. It is generally regarded as a universal catalyst of respiration, forming an essential electron-bridge between the respirable substrates and oxygen.
- Identity test: Passes
Cytochrome c is generally regarded as a universal catalyst of respiration, forming an essential electron-bridge between the respirable substrates and oxygen.
Dark red to red-brown solution at 10 mg plus 1 ml of water.
: Research Use Only (RUO).
- CAS No.:9007-43-6
- Source:Heart
- Species:Horse
- Form:Maroon powder
- MDL No.:MFCD00130890
- Protein/peptide name:Cytochrome C
- Purity:≥90%
- Molecular weight:884,893 g/mol
- Molecular formula:C₄₂H₅₂FeN₈O₆S₂