Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Environmental Test Kits
Powder pillows, aluminium
Powder pillows, aluminium
Powder pillows, aluminium
Catalog # HACH1429099
Supplier:
Hach
CAS Number:
Expand image
Powder pillows, aluminium
Catalog # HACH1429099
Supplier:
Hach
CAS Number:
Specifications
Chemical method:
Aluminon
Pk:
100
Description:
Aluminum (AluVer® 3)
Type:
PP
Range:
0 - 0,80 mg/L
Specifications
About this item
Product Details
Documentation
Product Details
Documentation