Cut resistant gloves, Cut 5 PU Palm, A622
Supplier: Portwest
Certificates
About this item
Level 5 cut resistant gloves for maximum protection against cuts and sharp objects. Made of HPPE/glass fibre fabric with a PU coating on palm and fingertips.
- PU dipped palm provides excellent grip
- Seamless liner for added comfort and dexterity
Certifications: EN 388: 2016 + A1: 2018 4X43C
Specifications
- Catalog No:
- PRTWA622G7RXL
- PRTWA622G7RL
- PRTWA622G7RXXL
- PRTWA622G7RM
- Model:
- Cut 5 PU Palm, A622
- Cut 5 PU Palm, A622
- Cut 5 PU Palm, A622
- Cut 5 PU Palm, A622
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Liner material:
- HPPE/glass fibre
- HPPE/glass fibre
- HPPE/glass fibre
- HPPE/glass fibre
- Finish:
- PU coating on palm and fingertips
- PU coating on palm and fingertips
- PU coating on palm and fingertips
- PU coating on palm and fingertips
- Gauge:
- 13
- 13
- 13
- 13
- Cuff style:
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- EN norm:
- EN 388: 2016 + A1: 2018 4X43C
- EN 388: 2016 + A1: 2018 4X43C
- EN 388: 2016 + A1: 2018 4X43C
- EN 388: 2016 + A1: 2018 4X43C