Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Colour Reference and Spectroscopy Standards
Primary colour solutions, Reag. Ph. Eur.
Primary colour solutions, Reag. Ph. Eur.
Primary colour solutions, Reag. Ph. Eur.
Supplier:
Ricca Chemical
Certificates
Expand image
Primary colour solutions, Reag. Ph. Eur.
Supplier:
Ricca Chemical
About this item
Related Information
Documentation
Certificates
Search by Lot Number
Documentation