Order Entry
Singapore
ContactUsLinkComponent
Val-Gly-Ser-Glu, MP Biomedicals
Val-Gly-Ser-Glu, MP Biomedicals
Catalog # ICNA0215277605
Supplier:  MP Biomedicals
CAS Number:  
Val-Gly-Ser-Glu, MP Biomedicals
Catalog # ICNA0215277605
Supplier:  MP Biomedicals
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Seq:
    02
  • Storage conditions:
    0 °C
  • Form:
    White Powder
  • Protein synonyms:
    VGSG|Eosinophilotactic Tetrapeptide
  • Protein/peptide name:
    Val-Gly-Ser-Glu

Specifications

About this item