Order Entry
Singapore
ContactUsLinkComponent
 
Wheat germ lectin, MP Biomedicals
 
Wheat germ lectin, MP Biomedicals

 

  • Species:
    Wheat germ
  • Storage conditions:
    −20 °C
  • Protein/peptide name:
    Lectin
  • Formulation:
    Lyophilised powder