
(R)-(+)-β-Hydroxy-γ-butyrolactone ≥90%, ee 99%
Catalog # ALFAL20255.06
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:102.089 g/mol
Boiling Pt:98…100 °C (0,3 mmHg)
Density:1,302
Flash Pt:>110 °C (230 °F)
:GHS07
CAS Number:58081-05-3
Formula:C₄H₆O₃
MDL Number:MFCD00211248