2,3,4-Tri-O-benzyl-β-D-arabinopyranose ≥97%
Catalog # ALFAL19870.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
- Environmentally Preferable:
Specifications
About this item
:
:420.51 g/mol
Melting Pt:73…75 °C
CAS Number:18039-26-4
Formula:C₂₆H₂₈O₅
MDL Number:MFCD05664045