Poly(styrene-divinylbenzene), aminomethylated 2,0-3,0 mmol/g on polystyrene
Catalog # ALFAL19471.06
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
About this item
CAS Number:89551-24-6
:
MDL Number:MFCD00130502
Storage Temperature:Ambient