Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Thiomorpholine, polymer supported
Thiomorpholine, polymer supported 0,8 - 1,1 mmol/g
Thiomorpholine, polymer supported 0,8 - 1,1 mmol/g
Thiomorpholine, polymer supported 0,8 - 1,1 mmol/g
Catalog # ALFAL19470.14
Supplier:
Thermo Fisher Scientific
CAS Number:
SDS
Expand image
Thiomorpholine, polymer supported 0,8 - 1,1 mmol/g
Catalog # ALFAL19470.14
Supplier:
Thermo Fisher Scientific
CAS Number:
SDS
Specifications
Pk:
25 g
Seq:
3308
Specifications
About this item
Product Details
Documentation
:
MDL Number:
MFCD03458715
Storage Temperature:
Ambient
Product Details
Documentation