Arbutin 98+% (by HPLC)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:497-76-7
:
Formula:C₁₂H₁₆O₇
MDL Number:MFCD00016915
:272.25 g/mol
Merck Index:14,773
Melting Pt:196…200 °C
Storage Temperature:Ambient
EINECS:207-850-3
Specification Test Results
- Appearance (Color):White
- Form:Powder
- Assay (HPLC):98.0%
- Optical Rotation:−65° ± 2° (c=2 in water)