Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Ethyl-3-(4-methoxybenzoyl)propionate
Ethyl-3-(4-methoxybenzoyl)propionate 98%
Ethyl-3-(4-methoxybenzoyl)propionate 98%
Ethyl-3-(4-methoxybenzoyl)propionate 98%
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
Ethyl-3-(4-methoxybenzoyl)propionate 98%
Supplier:
Thermo Fisher Scientific
About this item
CAS Number:
15118-67-9
:
Formula:
C₁₃H₁₆O₄
MDL Number:
MFCD00051784
:
236.27 g/mol
Melting Pt:
49…53 °C
Storage Temperature:
Ambient
Related Information
Documentation
SDS
msds_ALFAL07989
Application Notes
Organic Building Blocks and Intermediates
Videos
Thermo Scientific Acroseal 25 Years
Documentation