Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Hydroxy-n-octanoic acid
2-Hydroxy-n-octanoic acid ≥98%
2-Hydroxy-n-octanoic acid ≥98%
Irritant
2-Hydroxy-n-octanoic acid ≥98%
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
Irritant
2-Hydroxy-n-octanoic acid ≥98%
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
617-73-2
:
Formula:
C₈H₁₆O₃
MDL Number:
MFCD00014410
Melting Pt:
66…71 °C
:
160.21 g/mol
EINECS:
210-524-3
:
GHS07
Storage Temperature:
Ambient
Related Information
Documentation
Certificates
Search by Lot Number
SDS
msds_ALFAL02314
Application Notes
Organic Building Blocks and Intermediates
Videos
Thermo Scientific Acroseal 25 Years
Documentation