
Dextromethorphan hydrobromide ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:125-69-9
:
Formula:C₁₈H₂₆BrNO
:352.316 g/mol
Merck Index:14,8091
Melting Pt:125…127 °C
EINECS:204-750-1
:GHS07
Storage Temperature:Ambient