CHAPS lysis buffer (2X)
Supplier: Thermo Fisher Scientific
Certificates
About this item
Physical Form: Liquid
Appearance: Clear liquid
MDL No.: MFCD00282866
Specifications
- MDL No.:MFCD00282866
- Physical form:Liquid
- Appearance:Clear liquid
Physical Form: Liquid
Appearance: Clear liquid
MDL No.: MFCD00282866