Order Entry
Singapore
Orders LinkContactUsLinkComponent
 

Doxycycline hyclate

Doxycycline hyclate
Supplier:  
Certificates
Doxycycline hyclate
Doxycycline hyclate

0 Options Available Below

Product Details & Documents

CAS No.: 24390-14-5 Molecular Formula: C22H24N2O8.HCl.0.5H2O.0.5C2H6O Formula Weight: 512.94 (503.94anhy) Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: Pale yellow Solubility: Soluble in water to 50mg/ml. May require warming MDL No.: MFCD07357237

Applications: Inhibits matrix metalloproteinases at subantimicrobial doses

Synonyms:  Doxycycline Hydrochloride Hemiethanolate Hemihydrate
  • CAS Number:24390-14-5
  • Formula:C₂₂H₂₄N₂O₈·HCl·0.5H₂O·0.5C₂H₆O
  • MDL Number:MFCD07357237
  • Molecular Weight:512.94 g/mol (503.94 anhy)
  • Merck Index:14,3440
Product Family Options