Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
N'-Hydroxy-2-pyrazinecarboximidamide
N'-Hydroxy-2-pyrazinecarboximidamide 97%
N'-Hydroxy-2-pyrazinecarboximidamide 97%
Irritant
N'-Hydroxy-2-pyrazinecarboximidamide 97%
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
Irritant
N'-Hydroxy-2-pyrazinecarboximidamide 97%
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
51285-05-3
:
Formula:
C₅H₆N₄O
MDL Number:
MFCD02258397
:
138.13 g/mol
:
GHS07
Storage Temperature:
Ambient
Related Information
Documentation
Certificates
Search by Lot Number
SDS
msds_ALFAH54834
Application Notes
Organic Building Blocks and Intermediates
Videos
Thermo Scientific Acroseal 25 Years
Documentation