Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Pentynoic acid
2-Pentynoic acid 97%
2-Pentynoic acid 97%
Irritant
2-Pentynoic acid 97%
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
Irritant
2-Pentynoic acid 97%
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
5963-77-9
:
Formula:
C₅H₆O₂
MDL Number:
MFCD00014370
:
98.10 g/mol
Melting Pt:
47…53 °C
Flash Pt:
96 °C (205 °F)
:
GHS07
Storage Temperature:
Ambient
Related Information
Documentation
Certificates
Search by Lot Number
SDS
msds_ALFAH53411
Application Notes
Organic Building Blocks and Intermediates
Videos
Thermo Scientific Acroseal 25 Years
Documentation