Order Entry
Singapore
ContactUsLinkComponent
10-(1-Naphthyl)anthracene-9-boronic acid 97%
Irritant
10-(1-Naphthyl)anthracene-9-boronic acid 97%
Catalog # ALFAH52842.MD
CAS Number:  
Irritant
10-(1-Naphthyl)anthracene-9-boronic acid 97%
Catalog # ALFAH52842.MD
CAS Number:  

Specifications

  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :348.21 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:400607-46-7
Formula:C₂₄H₁₇BO₂
MDL Number:MFCD11977302