Order Entry
Singapore
ContactUsLinkComponent
D-β-Homoproline hydrochloride 95%
D-β-Homoproline hydrochloride 95%
Catalog # ALFAH52085.MD
CAS Number:  
D-β-Homoproline hydrochloride 95%
Catalog # ALFAH52085.MD
CAS Number:  

Specifications

  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

CAS Number:439918-59-9
 :
Formula:C₆H₁₁NO₂·HCl
MDL Number:MFCD06202401
 :165.62 g/mol
Melting Pt:195…197 °C
Storage Temperature:Ambient