Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Fmoc-beta-(3-benzothienyl)-Ala-OH
Fmoc-β-(3-benzothienyl)-Ala-OH 95%
Fmoc-β-(3-benzothienyl)-Ala-OH 95%
:
Thermo Fisher Scientific
Fmoc-β-(3-benzothienyl)-Ala-OH 95%
:
Thermo Fisher Scientific
:
177966-60-8
:
:
C₂₆H₂₁NO₄S
:
MFCD00672562
:
443.517 g/mol
msds_ALFAH52054
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years