Order Entry
Singapore
ContactUsLinkComponent
2-(1,3-Dimethyl-2,4,6-trioxohexahydropyrimidin-5-ylidenemethyl)thiophene-5-boronic acid pinacol ester 95%
2-(1,3-Dimethyl-2,4,6-trioxohexahydropyrimidin-5-ylidenemethyl)thiophene-5-boronic acid pinacol ester 95%
Catalog # ALFAH51898.06
CAS Number:  
2-(1,3-Dimethyl-2,4,6-trioxohexahydropyrimidin-5-ylidenemethyl)thiophene-5-boronic acid pinacol ester 95%
Catalog # ALFAH51898.06
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :376.23 g/mol
Storage Temperature:Ambient
Formula:C₁₇H₂₁BN₂O₅S
MDL Number:MFCD16294525