Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-(9-Anthryl)propionic acid
3-(9-Anthryl)propionic acid 96%
3-(9-Anthryl)propionic acid 96%
Irritant
3-(9-Anthryl)propionic acid 96%
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
Irritant
3-(9-Anthryl)propionic acid 96%
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
41034-83-7
:
Formula:
C₁₇H₁₄O₂
MDL Number:
MFCD00068641
Melting Pt:
193…197 °C
:
250.30 g/mol
:
GHS07
Storage Temperature:
Ambient
Related Information
Documentation
Certificates
Search by Lot Number
SDS
msds_ALFAH34184
Application Notes
Organic Building Blocks and Intermediates
Videos
Thermo Scientific Acroseal 25 Years
Documentation