Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-(3-Chlorophenylsulphonyl)-L-proline
1-(3-Chlorophenylsulphonyl)-L-proline 96%
1-(3-Chlorophenylsulphonyl)-L-proline 96%
ALFAH33849.03
:
Thermo Fisher Scientific
:
1-(3-Chlorophenylsulphonyl)-L-proline 96%
ALFAH33849.03
:
Thermo Fisher Scientific
:
Pk:
1 g
Seq:
3047
:
:
C₁₁H₁₂ClNO₄S
:
MFCD07433679
:
289.74 g/mol
:
85…86 °C
:
GHS07
:
Ambient