$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Singapore
ContactUsLinkComponent
4-Bromo-1-methyl-1H-pyrazole-3-carbaldehyde
4-Bromo-1-methyl-1H-pyrazole-3-carbaldehyde 97%
4-Bromo-1-methyl-1H-pyrazole-3-carbaldehyde 97%
:
Thermo Fisher Scientific
4-Bromo-1-methyl-1H-pyrazole-3-carbaldehyde 97%
:
Thermo Fisher Scientific
:
287917-96-8
:
:
C₅H₅BrN₂O
:
MFCD00103235
:
189.02 g/mol
:
69…71 °C
:
GHS07
:
Ambient
msds_ALFAH33484
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years