
3-(2-Furyl)-1H-pyrazole 97%
Catalog # ALFAH33472.06
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:134.14 g/mol
Melting Pt:102…104 °C
:GHS07
Storage Temperature:Ambient
CAS Number:32332-98-2
Formula:C₇H₆N₂O
MDL Number:MFCD00973923