$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Singapore
ContactUsLinkComponent
1-(4'-Methoxy-4-biphenylylsulphonyl)-L-proline
1-(4'-Methoxy-4-biphenylylsulphonyl)-L-proline 96%
1-(4'-Methoxy-4-biphenylylsulphonyl)-L-proline 96%
Irritant
1-(4'-Methoxy-4-biphenylylsulphonyl)-L-proline 96%
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
Irritant
1-(4'-Methoxy-4-biphenylylsulphonyl)-L-proline 96%
Supplier:
Thermo Fisher Scientific
Certificates
About this item
:
Formula:
C₁₈H₁₉NO₅S
MDL Number:
MFCD06409466
:
361.42 g/mol
Melting Pt:
153…159 °C
:
GHS07
Storage Temperature:
Ambient
Related Information
Documentation
Certificates
Search by Lot Number
SDS
msds_ALFAH32890
Videos
Thermo Scientific Acroseal 25 Years
Documentation