Order Entry
Singapore
ContactUsLinkComponent
3-Amino-1-(4-chlorophenyl)-1H-pyrazole 95%
Irritant
3-Amino-1-(4-chlorophenyl)-1H-pyrazole 95%
Catalog # ALFAH32252.MD
CAS Number:  
Irritant
3-Amino-1-(4-chlorophenyl)-1H-pyrazole 95%
Catalog # ALFAH32252.MD
CAS Number:  

Specifications

  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

CAS Number:66000-39-3
 :
Formula:C₉H₈ClN₃
MDL Number:MFCD09027932
 :193.64 g/mol
Melting Pt:107…108 °C
 :GHS07
Storage Temperature:Ambient