Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-Bromobenzoxazole-2-thiol
5-Bromobenzoxazole-2-thiol 97%
5-Bromobenzoxazole-2-thiol 97%
ALFAH32237.03
:
Thermo Fisher Scientific
:
5-Bromobenzoxazole-2-thiol 97%
ALFAH32237.03
:
Thermo Fisher Scientific
:
Pk:
1 g
Seq:
3047
:
439607-87-1
:
:
C₇H₄BrNOS
:
MFCD08447020
:
230.08 g/mol
:
280…283 °C
:
GHS05
:
GHS07