Order Entry
Singapore
ContactUsLinkComponent
1-(Phenylsulphonyl)-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrrolo[2,3-b]pyridine 95%
Irritant
1-(Phenylsulphonyl)-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrrolo[2,3-b]pyridine 95%
Catalog # ALFAH31505.MD
CAS Number:  
Irritant
1-(Phenylsulphonyl)-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrrolo[2,3-b]pyridine 95%
Catalog # ALFAH31505.MD
CAS Number:  

Specifications

  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

CAS Number:886547-94-0
 :
Formula:C₁₉H₂₁BN₂O₄S
 :384.27 g/mol
 :GHS07
Storage Temperature:Ambient
MDL Number:MFCD09999194