Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Singapore
ContactUsLinkComponent
Home
3-Methyl-4-nitro-1H-pyrazole
3-Methyl-4-nitro-1H-pyrazole 97%
3-Methyl-4-nitro-1H-pyrazole 97%
:
Thermo Fisher Scientific
3-Methyl-4-nitro-1H-pyrazole 97%
:
Thermo Fisher Scientific
:
5334-39-4
:
:
C4H5N3O2
:
MFCD00037864
:
325 °C
:
131…134 °C
:
127.10 g/mol
:
GHS07
:
Ambient
msds_ALFAH30860
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years