(2S,3aS,7aS)-2-Carboxyoctahydroindole 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: (2S,3aS,7aS)-Octahydro-1H-indole-2-carboxylic acid, H-Oic-OH
CAS Number:80875-98-5
:
Formula:C₉H₁₅NO₂
MDL Number:MFCD07782125
:169.22 g/mol
Melting Pt:266 °C (dec.)
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable: