Order Entry
Singapore
ContactUsLinkComponent
4-Benzyloxy-5-bromo-2-chloropyrimidine 97%
4-Benzyloxy-5-bromo-2-chloropyrimidine 97%
Catalog # ALFAH27706.03
CAS Number:  
4-Benzyloxy-5-bromo-2-chloropyrimidine 97%
Catalog # ALFAH27706.03
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

CAS Number:205672-19-1
 :
Formula:C₁₁H₈BrClN₂O
MDL Number:MFCD06798237
 :299.56 g/mol
Melting Pt:114…116 °C
Storage Temperature:Ambient