Order Entry
Singapore
ContactUsLinkComponent
1-Methyl-1H-pyrazole-5-boronic acid neopentyl glycol ester 95%
  ALFAH27703.03
 :  
1-Methyl-1H-pyrazole-5-boronic acid neopentyl glycol ester 95%
  ALFAH27703.03
 :  
  • Pk:
    1 g
  • Seq:
    3047
 :1044851-76-4
 :
 :C₉H₁₅BN₂O₂
 :MFCD07777079
 :194.04 g/mol
 :72…75 °C
 :Ambient