Order Entry
Singapore
ContactUsLinkComponent
6,7-Dimethoxy-1,4-dihydro-2,3-quinoxalinedione monohydrate 98%
Irritant
6,7-Dimethoxy-1,4-dihydro-2,3-quinoxalinedione monohydrate 98%
Catalog # ALFAH25941.06
CAS Number:  
Irritant
6,7-Dimethoxy-1,4-dihydro-2,3-quinoxalinedione monohydrate 98%
Catalog # ALFAH25941.06
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

CAS Number:4784-02-5
 :
Formula:C₁₀H₁₀N₂O₄·H₂O
MDL Number:MFCD09953682
 :240.22 g/mol (222.20 anhy)
Melting Pt:352…354 °C
 :GHS07
Storage Temperature:Ambient