

tert-Butyl trichloroacetimidate 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:98946-18-0
:
Formula:C₆H₁₀Cl₃NO
MDL Number:MFCD00077410
:218.502 g/mol
Boiling Pt:64…66 °C (15 mmHg)
Melting Pt:20…22 °C
Density:1,222
Flash Pt:55 °C (131 °F)
ADR:3,III
:GHS02
:GHS07