Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
(1R)-(+)-2,10,Camphorsultam
(1R)-(+)-2,10,Camphorsultam 99%
(1R)-(+)-2,10,Camphorsultam 99%
Irritant
(1R)-(+)-2,10,Camphorsultam 99%
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
Irritant
(1R)-(+)-2,10,Camphorsultam 99%
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
108448-77-7
:
Formula:
C₁₀H₁₇NO₂S
MDL Number:
MFCD00151762
:
215.32 g/mol
Melting Pt:
179…186 °C
:
GHS07
Storage Temperature:
Ambient
Related Information
Documentation
Certificates
Search by Lot Number
SDS
msds_ALFAA14133
Application Notes
Organic Building Blocks and Intermediates
Videos
Thermo Scientific Acroseal 25 Years
Documentation