Silicone oil high temperature , colourless liquid for oil baths
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:68083-14-7
:63148-62-9
Formula:[Si(CH3)2O]x[Si(C6H5)2O]k
MDL Number:MFCD00132673
Merck Index:14,8495
Density:1,07 g/mL
Flash Pt:>300 °C(572 °F)