Order Entry
Singapore
ContactUsLinkComponent
Ethanol denatured ≥91.6% (v/v) (denatured with 3.7% MeOH, 1.9% MIBK, 1% each of heptane, ethylacetate and toluene )
Flammable
Health
Ethanol denatured ≥91.6% (v/v) (denatured with 3.7% MeOH, 1.9% MIBK, 1% each of heptane, ethylacetate and toluene )
Catalog # ALFA45844.K2
CAS Number:  
Flammable
Health
Ethanol denatured ≥91.6% (v/v) (denatured with 3.7% MeOH, 1.9% MIBK, 1% each of heptane, ethylacetate and toluene )
Catalog # ALFA45844.K2
CAS Number:  

Specifications

  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :46.07 g/mol
Merck Index:14,3760
Boiling Pt:78 °C
Melting Pt:–130 °C
Density:0,789
Flash Pt:8 °C (46 °F)
Storage Temperature:Ambient
EINECS:200-578-6
UN:1170
ADR:3,II
 :GHS02
 :GHS08
CAS Number:64-17-5
Formula:H₃CCH₂OH
MDL Number:MFCD00003568