$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Singapore
ContactUsLinkComponent
m-Carborane
m-Carborane ≥98%
m-Carborane ≥98%
ALFA45606.ME
:
Thermo Fisher Scientific
:
m-Carborane ≥98%
ALFA45606.ME
:
Thermo Fisher Scientific
:
Pk:
500 mg
Seq:
2725
:
:
144.23 g/mol
:
GHS07
:
Ambient
:
16986-24-6
:
C₂H₁₂B₁₀
:
MFCD00151574