Order Entry
Singapore
Orders LinkContactUsLinkComponent
Malic enzyme

Malic enzyme

1 KU
Catalog # ALFA45396.4I
Malic enzyme
Catalog # ALFA45396.4I
CAS Number:  
Malic enzyme
Malic enzyme

1 KU

CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 1 KU
Product Details & Documents

Applications: Cofactor regeneration of NADH in reductive processes. Diagnostics

Specifications

Specifications

  • Pk:
    1 KU
  • Cat. No.:
    ALFA45396.4I
  • Form:
    White powder
  • MDL No.:
    MFCD00131568
  • Enzyme name:
    Malic Enzyme

Specifications