Order Entry
Singapore
ContactUsLinkComponent
Palladium(II) chloride solution (20 - 25% Pd) (w/w)
Corrosive
Irritant
Palladium(II) chloride solution (20 - 25% Pd) (w/w)
Catalog # ALFA40019.NH
CAS Number:  
Corrosive
Irritant
Palladium(II) chloride solution (20 - 25% Pd) (w/w)
Catalog # ALFA40019.NH
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
ADR:8,III
 :177.33 g/mol
EINECS:231-596-2
UN:3264
 :GHS05
 :GHS07
Storage Temperature:Ambient
CAS Number:7647-10-1
Formula:PdCl₂
MDL Number:MFCD00003558