Order Entry
Singapore
Orders LinkContactUsLinkComponent
Multibore insulating tubes, ceramic
Multibore insulating tubes, ceramic
Certificates
Multibore insulating tubes, ceramic
3 Options Available Below
Product Details & Documents
Pure Al₂O₃ Aluminium oxide, AL23.
Dense, excellent thermal and electrical resistance properties, corrosion resistant, permeable to microwaves.
Product Family Options
Product Information
  • Tubing int.Ø
    Tubing ext.Ø
    Length
    Availability
    Price
  • 4 Bores
    1,2 mm
    5,5 mm
    1460 mm
    Log in for availability
    Log in for availability
    1 * 1.460 mm
    1 * 1.460 mm
  • 4 Bores
    1,5 mm
    8,5 mm
    380 mm
    Log in for availability
    Log in for availability
    1 * 380 mm
    1 * 380 mm
  • 2 Bores
    1,6 mm
    8,0 mm
    770 mm
    Log in for availability
    Log in for availability
    1 * 770 mm
    1 * 770 mm