Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Rhodium (III) sulphate
Rhodium(III) sulphate (5% Rh) bright plating solution
Rhodium(III) sulphate (5% Rh) bright plating solution
ALFA12635.NJ
:
Thermo Fisher Scientific
:
Rhodium(III) sulphate (5% Rh) bright plating solution
ALFA12635.NJ
:
Thermo Fisher Scientific
:
Pk:
0,5 g
Seq:
3035
Environmentally Preferable:
:
Dirhodium trisulphate
:
:
494
:
234-014-5
:
3264
:
GHS05
:
Ambient
:
10489-46-0
:
O₁₂Rh₂S₃
:
MFCD00016309
:
1.4
:
8 III