Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Chromium(III) acetate hydroxide
Chromium(III) acetate hydroxide (24% Cr)
Chromium(III) acetate hydroxide (24% Cr)
Chromium(III) acetate hydroxide (24% Cr)
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
Chromium(III) acetate hydroxide (24% Cr)
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
39430-51-8
:
Formula:
C₁₄H₂₃Cr₃O₁₆
MDL Number:
MFCD00058912
:
603.311 g/mol
EINECS:
254-447-3
:
GHS07
Storage Temperature:
Ambient
Related Information
Documentation
Certificates
Search by Lot Number
SDS
msds_ALFA012518
Videos
Thermo Scientific Acroseal 25 Years
Documentation