Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Stainless steel
Stainless steel (Fe:Cr:Ni; 70:19:11 wt%), powder -100 mesh Type 304-L
Stainless steel (Fe:Cr:Ni; 70:19:11 wt%), powder -100 mesh Type 304-L
Stainless steel (Fe:Cr:Ni; 70:19:11 wt%), powder -100 mesh Type 304-L
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
Stainless steel (Fe:Cr:Ni; 70:19:11 wt%), powder -100 mesh Type 304-L
Supplier:
Thermo Fisher Scientific
Certificates
About this item
:
Formula:
Fe
MDL Number:
MFCD00148499
:
55.845 g/mol
Storage Temperature:
Ambient
Related Information
Documentation
Certificates
Search by Lot Number
SDS
msds_ALFA011085
Videos
Thermo Scientific Acroseal 25 Years
Documentation