Order Entry
Singapore
ContactUsLinkComponent
Electro-blotting systems, omniPAGE Mini
Electro-blotting systems, omniPAGE Mini
Supplier:  CLEAVER SCIENTIFIC
Certificates
undefined
Electro-blotting systems, omniPAGE Mini
Supplier:  CLEAVER SCIENTIFIC
Specifications
  • Catalog No:
  • CLEVCVS10CBS-HI
  • CLEVCVS10CBS
  • Model:
  • omniPAGE mini
  • omniPAGE mini
  • Buffer volume:
  • 1200 ml
  • 1200 ml
  • Capacity:
  • 4 gels/transfer
  • 4 gels/transfer
  • Distance between electrodes:
  • 80 mm
  • 80 mm
Specifications