Trisodium 2-(4-Sulphophenylazo)-1,8-dihydroxynaphthalene-3,6-disulphonate (SPADNS), BAKER ANALYZED® indicator
Catalog # BAKRJ189-02
Supplier: Avantor
CAS Number:
Specifications
- Pack type:Glass bottle, wide mouth, amber, black T/E cap
- Pk:10 g
- Seq:3236
Specifications
Specification Test Results
- Sensitivity as Indicator:Passes test
Specification Test Results
About this item
:
:570.40 g/mol
CAS Number:23647-14-5
Formula:C₁₆H₁₂N₂O₁₁S₃
MDL Number:MFCD00003952